Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNDP1 Monoclonal Antibody (CL0339)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531348
Description
Immunogen sequence: PALLEKVFQY IDLHQDEFVQ TLKEWVAIES DSVQPVPRFR QELFRMMAVA ADTLQRLGAR VASVDMGPQQ LPDGQSLPIP PVILAELGSD PTKGTVCFYG HL.
This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
Specifications
CNDP1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96KN2 | |
CNDP1 | |
Recombinant protein corresponding to Human CNDP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
CL0339 | |
Unconjugated | |
CNDP1 | |
AI746433; beta-Ala-His dipeptidase; carnosinase 1; carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CN1; CNDP dipeptidase 1; CNDP1; CPGL2; glutamate carboxypeptidase-like protein 2; HsT2308; serum carnosinase; UNQ1915/PRO4380 | |
Mouse | |
Protein A | |
RUO | |
84735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction