Learn More
CNDP1 Monoclonal Antibody (OTI2D2), TrueMAB™, OriGene
SDP
Mouse Monoclonal Antibody
Supplier:  OriGene TA502093
Encompass_Preferred
 View more versions of this product
Catalog No. 50-167-3177
Only null left
Add to Cart
Description
Description
This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
Specifications
Specifications
CNDP1
Monoclonal
Unconjugated
CNDP1
CN1, CPGL2, HsT2308
Mouse
Affinity Chromatography
RUO
84735
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI2D2
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96KN2
CNDP1
Full length human recombit protein of human CNDP1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.