Learn More
CNDP1 Mouse anti-Human, Clone: OTI2D2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502093

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.Specifications
CNDP1 | |
Monoclonal | |
Unconjugated | |
CNDP1 | |
CN1, CPGL2, HsT2308 | |
Mouse | |
Affinity Chromatography | |
RUO | |
84735 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2D2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96KN2 | |
CNDP1 | |
Full length human recombit protein of human CNDP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.