Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNDP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121107
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121107 Supplier Invitrogen™ Supplier No. PA5121107
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, A-431, HepG2, Mouse kidney, Mouse brain, Rat spinal cord, Rat brain. The target is usually found in the following locations: Secreted. Immunogen sequence: MDPKLGRMAA SLLAVLLLLL ERGMFSSPSP PPALLEKVFQ YIDLHQDEFV QTLKEWVAIE SDSVQPVPRF RQELFRMMAV AADTLQRLGA RVASVDMGPQ QLPDGQSLPI PPVILAELGS DPTKGTVCFY GHLDVQPADR GDGWLTDPYV LTEVDGKLYG RGATDNKGPV LAWINAVSAF RALEQDLPVN IKFIIEGMEE AGSVALEELV EKEKDRFFSG VDYIVISDNL WISQRKPAIT YGTRGNSYFM VEVKCRDQDF HSGTFGGILH EPMADLVALL.

This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNDP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CNDP1
Gene Accession No. Q66HG3, Q8BUG2, Q96KN2
Gene Alias AI746433; beta-Ala-His dipeptidase; carnosinase 1; carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CN1; CNDP dipeptidase 1; CNDP1; CPGL2; glutamate carboxypeptidase-like protein 2; HsT2308; serum carnosinase; UNQ1915/PRO4380
Gene Symbols CNDP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CNDP1 (NP_116038.4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307212, 338403, 84735
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.