Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNDP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528965 Supplier Invitrogen™ Supplier No. PA528965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (83%), Rat (82%), Pig (85%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNDP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CNDP1
Gene Accession No. Q96KN2
Gene Alias AI746433; beta-Ala-His dipeptidase; carnosinase 1; carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CN1; CNDP dipeptidase 1; CNDP1; CPGL2; glutamate carboxypeptidase-like protein 2; HsT2308; serum carnosinase; UNQ1915/PRO4380
Gene Symbols CNDP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CNDP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84735
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.