Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNDP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547332
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547332 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547332 Supplier Invitrogen™ Supplier No. PA547332
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNDP1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CNDP1
Gene Accession No. Q96KN2
Gene Alias AI746433; beta-Ala-His dipeptidase; carnosinase 1; carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CN1; CNDP dipeptidase 1; CNDP1; CPGL2; glutamate carboxypeptidase-like protein 2; HsT2308; serum carnosinase; UNQ1915/PRO4380
Gene Symbols CNDP1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carnosine Dipeptidase 1/CNDP1 Pro28-His507.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84735
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.