Learn More
Invitrogen™ CNDP2 Monoclonal Antibody (464702)

Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human CNDP1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CNDP2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 464702 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Cndp2 |
| Gene Accession No. | Q96KP4 |
| Gene Alias | 0610010E05Rik; C76600; carnosine dipeptidase 2; carnosine dipeptidase II; CN2; CNDP dipeptidase 2; CNDP dipeptidase 2 (metallopeptidase M20 family); Cndp2; CPGL; cytosolic nonspecific dipeptidase; Cytosolic non-specific dipeptidase; cytosolic nonspecific dipeptidase 2; Dip-2; Epididymis secretory protein Li 13; glutamate carboxypeptidase-like protein 1; HEL-S-13; HsT2298; Pep1; Pep-1; PEPA; peptidase 1; peptidase A; wu:fd20d11; wu:fe17f09; zgc:92569 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.