Learn More
CNDP2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305566A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
CNDP2, also known as tissue carnosinase and peptidase A (EC 3.4.13.18), is a nonspecific dipeptidase rather than a selective carnosinase (Teufel et al., 2003 [PubMed 12473676]).Specifications
CNDP2 | |
Polyclonal | |
Unconjugated | |
CNDP2 | |
CN2, CPGL, HsT2298, PEPA, cytosolic non-specific dipeptidase, cytosolic nonspecific dipeptidase, glutamate carboxypeptidase-like protein 1, peptidase A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55748 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96KP4 | |
CNDP2 | |
Between 425 and 475 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.