Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNEP1R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560136 Supplier Invitrogen™ Supplier No. PA560136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNSLEQAEDL KAFERRLTEY IHCLQPATGR WRSVLLHII Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 85%.

Forms with the serine/threonine protein phosphatase CTDNEP1 an active complex which dephosphorylates and may activate LPIN1 and LPIN2. LPIN1 and LPIN2 are phosphatidate phosphatases that catalyze the conversion of phosphatidic acid to diacylglycerol and control the metabolism of fatty acids at differents levels. May indirectly modulate the lipid composition of nuclear and/or endoplasmic reticulum membranes and be required for proper nuclear membrane morphology and/or dynamics. May also indirectly regulate the production of lipid droplets and triacylglycerol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNEP1R1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNEP1R1
Gene Accession No. Q8N9A8
Gene Alias C16orf69; CNEP1R1; CTD nuclear envelope phosphatase 1 regulatory subunit 1; NEP1R1; NEP1-R1; nuclear envelope phosphatase 1-regulatory subunit 1; nuclear envelope phosphatase-regulatory subunit 1; TMEM188; TMP125; Transmembrane protein 188
Gene Symbols CNEP1R1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNEP1R1. Recombinant protein control fragment (Product #RP-98804).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255919
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.