Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNG1 Antibody (CNC 9C1), Novus Biologicals™
SDP

Catalog No. NB10074389 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074389 Supplier Novus Biologicals Supplier No. NB10074389
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CNG1 Monoclonal specifically detects CNG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CNG1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CNC 9C1
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:100
Formulation Ascites with 0.05% Sodium Azide
Gene Alias cGMP-gated cation channel alpha-1, CNCG, CNCG1RP49, CNG-1, CNG1CNG channel alpha-1, cyclic nucleotide gated channel alpha 1, interleukin-1 homologue, photoreceptor, RCNC1, Rod photoreceptor cGMP-gated channel subunit alpha
Gene Symbols CNGA1
Host Species Mouse
Immunogen MBP-fusion protein of the C-terminal region of the a2 subunit (amino acids K581-D670)
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1259
Test Specificity CNG Channel (CNC 9C1)
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.