Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNGA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305204
Description
CNGA2 Polyclonal specifically detects CNGA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CNGA2 | |
Polyclonal | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:10-1:50, Immunohistochemistry-Paraffin | |
CNCA, CNCA1, CNCG2, CNG2CNG-2, cyclic nucleotide gated channel alpha 2, Cyclic nucleotide-gated cation channel 2, Cyclic nucleotide-gated channel alpha-2, cyclic nucleotide-gated olfactory channel, cyclic-nucleotide-gated cation channel 2, cyclic-nucleotide-gated olfactory channel, FLJ46312, OCNC1CNG channel alpha-2, OCNCa, OCNCalpha | |
Recombinant protein of human CNGA2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS with 50% glycerol, pH7.3. with 0.05% Proclin 300 | |
Rabbit | |
Affinity purified | |
RUO | |
1260 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction