Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNGA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522235 Supplier Invitrogen™ Supplier No. PA522235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, mouse brain. Predicted reactivity: Mouse (97%), Rat (96%), Rabbit (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CNGA2 represents the alpha subunit of a cyclic nucleotide-gated olfactory channel. The encoded protein contains a carboxy-terminal leucine zipper that mediates channel formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNGA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CNGA2
Gene Accession No. Q16280, Q62398
Gene Alias Cnca; CNCA1; CNCG2; Cncg4; cng channel; CNG channel 2; CNG channel alpha-2; CNG2; CNG-2; Cnga2; cyclic nucleotide gated channel 4; cyclic nucleotide gated channel alpha 2; cyclic nucleotide-gated cation channel 2; Cyclic nucleotide-gated channel alpha-2; cyclic nucleotide-gated olfactory channel; Cyclic nucleotide-gated olfactory channel subunit OCNC1; cyclic-nucleotide-gated cation channel 2; OCNC1; OCNCa; OCNCALPHA; OLFCH
Gene Symbols CNGA2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human CNGA2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1260, 12789
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.