Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNGA2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550017

Catalog No. PA550017


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total CNGA2 protein.

CNGA2 represents the alpha subunit of a cyclic nucleotide-gated olfactory channel. The encoded protein contains a carboxy-terminal leucine zipper that mediates channel formation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CNGA2
Polyclonal
Unconjugated
CNGA2
Cnca; CNCA1; CNCG2; Cncg4; cng channel; CNG channel 2; CNG channel alpha-2; CNG2; CNG-2; Cnga2; cyclic nucleotide gated channel 4; cyclic nucleotide gated channel alpha 2; cyclic nucleotide-gated cation channel 2; Cyclic nucleotide-gated channel alpha-2; cyclic nucleotide-gated olfactory channel; Cyclic nucleotide-gated olfactory channel subunit OCNC1; cyclic-nucleotide-gated cation channel 2; OCNC1; OCNCa; OCNCALPHA; OLFCH
Rabbit
Antigen Affinity Chromatography
RUO
1260, 12789
-20°C
Liquid
Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q16280, Q62398
CNGA2
Synthesized peptide derived from internal of human CNGA2 (aa 391-440).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.