Learn More
Invitrogen™ CNGA2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total CNGA2.
Specifications
Specifications
| Antigen | CNGA2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | CNGA2 |
| Gene Accession No. | Q00195, Q16280, Q62398 |
| Gene Alias | Cnca; CNCA1; CNCG2; Cncg4; cng channel; CNG channel 2; CNG channel alpha-2; CNG2; CNG-2; Cnga2; cyclic nucleotide gated channel 4; cyclic nucleotide gated channel alpha 2; cyclic nucleotide-gated cation channel 2; Cyclic nucleotide-gated channel alpha-2; cyclic nucleotide-gated olfactory channel; Cyclic nucleotide-gated olfactory channel subunit OCNC1; cyclic-nucleotide-gated cation channel 2; OCNC1; OCNCa; OCNCALPHA; OLFCH |
| Gene Symbols | CNGA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.