Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNGA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550211 Supplier Invitrogen™ Supplier No. PA550211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CNGA3 protein.

This gene encodes a member of the cyclic nucleotide-gated cation channel protein family which is required for normal vision and olfactory signal transduction. Mutations in this gene are associated with achromatopsia (rod monochromacy) and color blindness. Two alternatively spliced transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNGA3
Applications Western Blot
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CNGA3
Gene Accession No. Q16281, Q9JJZ8
Gene Alias ACHM2; alpha subunit of cone photoreceptor CNG-channel; CCNC1; CCNCa; CCNCalpha; CNCG3; cng channel; CNG channel 1; CNG channel alpha-3; CNG1; CNG-1; CNG3; CNG-3; CNGA3; CNGalpha2; cone photoreceptor cGMP-gated channel alpha subunit; cone photoreceptor cGMP-gated channel subunit alpha; cyclic nucleotide gated channel alpha 3; cyclic nucleotide-gated cation channel alpha-3; Cyclic nucleotide-gated channel alpha-3; cyclic nucleotide-gated channel cone photoreceptor subunit alpha
Gene Symbols CNGA3
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human cyclic nucleotide gated channel alpha 3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1261, 12790
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.