Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNGA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114327
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114327 Supplier Invitrogen™ Supplier No. PA5114327
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNGA4 Polyclonal Antibody for Flow, ICC/IF, Western Blot

Second messenger, cAMP, causes the opening of cation- selective cyclic nucleotide-gated (CNG) channels and depolarization of the neuron (olfactory sensory neurons, OSNs). CNGA4 is the modulatory subunit of this channel which is known to play a central role in the transduction of odorant signals and subsequent adaptation. By accelerating the calcium-mediated negative feedback in olfactory signaling it allows rapid adaptation to odor stimulation and extends its range of odor detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNGA4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CNGA4
Gene Accession No. Q3UW12, Q64359, Q8IV77
Gene Alias Cgn2; CNCA2; CNCalpha4, CNG5; cng channel; CNG channel alpha-4; CNG4; CNG-4; CNG5; CNGA4; CNGB2; cyclic nucleotide gated cation channel; cyclic nucleotide gated channel alpha 4; cyclic nucleotide gated channel beta 2; cyclic nucleotide-gated cation channel alpha 4; cyclic nucleotide-gated cation channel alpha-4; Cyclic nucleotide-gated channel alpha-4; cyclic nucleotide-gated olfactory channel subunit OCNC2; OCNC2; OCNCb; OCNCBETA; olfactory cyclic nucleotide-gated channel; rOCNC2
Gene Symbols CNGA4
Host Species Rabbit
Immunogen Synthetic peptide within rat CNGA4 aa 1-50.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1262, 233649, 85258
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.