Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNGB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112936
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112936 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112936 Supplier Invitrogen™ Supplier No. PA5112936
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNGB1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Glutamic acid rich protein (GARP) is a soluble protein localized to the outer segments of the rod photoreceptor. GARPs are characterized by six repeated motifs of: V V E K K N/E E. These repeat motifs help form an alpha-helix structure. The function(s) of GARP have not been fully characterized, but studies have suggested that GARP helps limit cGMP turnover by inhibiting phosphodiesterase activity. To date, there have been two splice variants characterized: GARP1 (full-length GARP) and GARP2 (truncated GARP). GARP also interacts with part of the larger cytoplasmic domain of the beta-subunit of the cyclic GMP-gated channel found in the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNGB1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CNGB1
Gene Accession No. Q14028
Gene Alias 240 kDa protein of rod photoreceptor CNG-channel; 240K protein of rod photoreceptor cng-channel; CNCG2; CNCG3L; CNCG4; cng channel; CNG channel 4; CNG channel beta-1; CNG4; CNG-4; CNG4c protein; CNGB1; CNGB1B; cyclic nucleotide gated channel beta 1; cyclic nucleotide gated channel subunit beta 1; cyclic nucleotide-gated cation channel 4; Cyclic nucleotide-gated cation channel beta-1; cyclic nucleotide-gated cation channel gamma; Cyclic nucleotide-gated cation channel modulatory subunit; cyclic nucleotide-gated channel beta subunit 1; cyclic nucleotide-gated channel beta subunit 1c; cyclic nucleotide-gated channel beta subunit 1d; cyclic nucleotide-gated channel beta subunit 1e; cyclic nucleotide-gated channel beta-1; GAR1; GARP; GARP2; glutamic acid-rich protein; glutamic-acid-rich protein; Gm1959; RCNC2; RCNCb; RCNCbeta; RP45; splice variant; tGARP; glutamic acid rich protein; truncated rod photoreceptor cGMP-gated channel beta-subunit
Gene Symbols CNGB1
Host Species Rabbit
Immunogen Recombinant Human Cyclic nucleotide-gated cation channel beta-1 protein (131-253AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1258
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.