Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNIH2/3 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8027R

Description
Involved in the transport and maturation of proteins.Specifications
CNIH2/3 | |
Polyclonal | |
Unconjugated | |
CNIH2 | |
CNIH2; CNIH-2; CNIL; Cornichon family AMPA receptor auxiliary protein 2; cornichon homolog 2; Cornichon-like protein; Protein cornichon homolog 2 | |
Rabbit | |
Protein A | |
RUO | |
12794, 254263, 361705 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O35089, Q5BJU5, Q6PI25 | |
CNIH2 | |
KLH conjugated synthetic peptide derived from human CNIH2/CNIH3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction