Learn More
CNIH2/CNIH3 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111788
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: RARERLKNIE RIC Homology: CNIH2 : Mouse, human identical.CNIH3 : Rat, mouse, human - 11/13 amino acid residues identical.
Cornichon homologs (CNIHs) are a family of AMPA-type glutamate receptor (AMPAR) auxiliary subunits that modulate AMPAR ion channel function and trafficking. Cornichon 2 and 3 (CNIH2/3) are responsible for controlling the export of AMPARs from the endoplasmic reticulum and associate with synaptic AMPARs to modulate channel kinetics. CNIHs are small proteins with three transmembrane domains that directly binds to AMPARs. Overexpression CNIHs and AMPA receptors causes their localization to the Golgi apparatus. CNIH and TARP both compete for the bindingto AMPAR subunitsGluA2,GluA3, andGluA4subunits suggesting both proteins bind to the same site. CNIH2/3 binding to AMPARs is dependent on AMPAR subunit composition and TARPs.
Specifications
CNIH2/CNIH3 (extracellular) | |
Polyclonal | |
Unconjugated | |
Cnih2 | |
2900075G08Rik; Cnih2; CNIH-2; Cnih3; CNIH-3; Cnil; Cornichon family AMPA receptor auxiliary protein 2; Cornichon family AMPA receptor auxiliary protein 3; Cornichon-like protein; Protein cornichon homolog 2; Protein cornichon homolog 3; RGD1304930 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12794, 361705, 690252, 72978 | |
-20°C | |
Lyophilized |
Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
D0Q0Y7, O35089, Q5BJU5, Q6ZWS4 | |
Cnih2, Cnih3 | |
Peptide corresponding to amino acid residues 49-61 of rat CNIH2 (Extracellular loop) | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.