Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNIH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113040
Description
CNIH2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Involved in the transport and maturation of proteins.
Specifications
CNIH2 | |
Polyclonal | |
Unconjugated | |
CNIH2 | |
Cnih2; CNIH-2; Cnih3; Cnil; Cornichon family AMPA receptor auxiliary protein 2; cornichon homolog 2; cornichon homolog 2 (Drosophila); cornichon homolog 3; cornichon-like protein; Protein cornichon homolog 2; RGD1304930 | |
Rabbit | |
Protein G | |
RUO | |
12794, 254263 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O35089, Q6PI25 | |
CNIH2 | |
Recombinant Human Protein cornichon homolog 2 protein (94-138AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction