Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNIH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535051
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535051 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535051 Supplier Invitrogen™ Supplier No. PA535051
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with chicken, mouse and rat based on sequence homology.

Involved in the transport and maturation of proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNIH2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CNIH2
Gene Accession No. Q6PI25
Gene Alias Cnih2; CNIH-2; Cnih3; Cnil; Cornichon family AMPA receptor auxiliary protein 2; cornichon homolog 2; cornichon homolog 2 (Drosophila); cornichon homolog 3; cornichon-like protein; Protein cornichon homolog 2; RGD1304930
Gene Symbols CNIH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 31-59 amino acids from the N-terminal region of human CNIH2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254263
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.