Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNIH3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548872
Description
Predicted to react with mouse and rat based on sequence homology.
Regulates the trafficking and gating properties of AMPA-selective glutamate receptors (AMPARs). Promotes their targeting to the cell membrane and synapses and modulates their gating properties by regulating their rates of activation, deactivation and desensitization. [UniProt].
Specifications
CNIH3 | |
Polyclonal | |
Unconjugated | |
CNIH3 | |
2900075G08Rik; Cnih3; CNIH-3; cornichon 3; cornichon family AMPA receptor auxiliary protein 3; cornichon homolog 3; cornichon homolog 3 (Drosophila); protein cornichon homolog 3 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
149111 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8TBE1 | |
CNIH3 | |
KLH conjugated synthetic peptide between 17-44 amino acids from the N-terminal region of human CNIH3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction