Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNIH4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113469
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113469 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113469 Supplier Invitrogen™ Supplier No. PA5113469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNIH4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CNIH4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CNIH4
Gene Accession No. Q9P003
Gene Alias CNIH4; CNIH-4; Cornichon family AMPA receptor auxiliary protein 4; cornichon homolog 4; HSPC163; Protein cornichon homolog 4
Gene Symbols CNIH4
Host Species Rabbit
Immunogen Recombinant Human Protein cornichon homolog 4 protein (78-117AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29097
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.