Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNKSR1 Mouse anti-Human, Clone: 1E11H8, Proteintech
SDP

Catalog No. 501737196 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7196 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7196 Supplier Proteintech Group Inc Supplier No. 667961IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a protein containing several motifs involved in protein-protein interaction, including PDZ, PH (Pleckstrin homology), and SAM (sterile alpha motif) domains. The encoded protein acts as a scaffold component for receptor tyrosine kinase signaling and may mediate crosstalk between different signaling pathways. Alternative splicing results in multiple transcript variants.

Specifications

Antigen CNKSR1
Applications Western Blot
Classification Monoclonal
Clone 1E11H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CNKSR1
Gene Accession No. Q969H4
Gene Alias CNK, CNK homolog protein 1, CNK1, CNKSR1, Connector enhancer of KSR 1, Connector enhancer of KSR like, hCNK1, KSR
Gene Symbols CNKSR1
Host Species Mouse
Immunogen CNKSR1 Fusion Protein Ag27855
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10256
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.