Learn More
Invitrogen™ CNKSR1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total CNKSR1 protein.
Specifications
Specifications
| Antigen | CNKSR1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CNKSR1 |
| Gene Accession No. | Q969H4 |
| Gene Alias | BC037758; CNK; CNK homolog protein 1; CNK1; CNKSR1; Connector enhancer of kinase suppressor of ras 1; connector enhancer of KSR 1; Connector enhancer of KSR-like |
| Gene Symbols | CNKSR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.