Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNKSR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556574 Supplier Invitrogen™ Supplier No. PA556574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPRTSFGSLT DSSEEALEGM VRGLRQGGVS LLGQPQPLTQ EQWRSSFMRR NRDPQLNERV HRVRALQSTL KAKLQELQVL EEVLGDPELT GEKFRQWKEQ NRE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 90%.

This gene encodes a protein containing several motifs involved in protein-protein interaction, including PDZ, PH (Pleckstrin homology), and SAM (sterile alpha motif) domains. The encoded protein acts as a scaffold component for receptor tyrosine kinase signaling and may mediate crosstalk between different signaling pathways. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNKSR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNKSR1
Gene Accession No. Q969H4
Gene Alias BC037758; CNK; CNK homolog protein 1; CNK1; CNKSR1; Connector enhancer of kinase suppressor of ras 1; connector enhancer of KSR 1; Connector enhancer of KSR-like
Gene Symbols CNKSR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNKSR1. Recombinant protein control fragment (Product #RP-94258).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10256
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.