Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNKSR2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988214
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8214 Supplier Bioss Supplier No. BS8029R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a multidomain protein that functions as a scaffold protein to mediate the mitogen-activated protein kinase pathways downstream from Ras. This gene product is induced by vitamin D and inhibits apoptosis in certain cancer cells. It may also play a role in ternary complex assembly of synaptic proteins at the postsynaptic membrane and coupling of signal transduction to membrane/cytoskeletal remodeling. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Dec 2009].

Specifications

Antigen CNKSR2
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CNKSR2
Gene Accession No. Q8WXI2
Gene Alias C030004A01; CNK homolog protein 2; CNK2; CNK2B; CNKSR2; Connector enhancer of kinase suppressor of ras 2; Connector enhancer of KSR 2; connector enhancer of KSR2; Kiaa0902; KSR2; KSR2B; Maguin; maguin-1; maguin-2; Membrane-associated guanylate kinase-interacting protein; mKIAA0902
Gene Symbols CNKSR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CNKSR2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22866
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.