Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNKSR3 Antibody (OTI1D7), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272428V
Description
CNKSR3 Monoclonal specifically detects CNKSR3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CNKSR3 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CNKSR3 | |
Full length recombinant protein of human CNKSR3 (NP_775786) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1D7 | |
Western Blot, Immunohistochemistry | |
CNK homolog protein 3, CNK3, CNKSR family member 3MAGI1, FLJ31349, membrane associated guanylate kinase interacting protein-like 1, membrane-associated guanylate kinase-interacting protein-like 1, WW and PDZ domain containing 1 | |
Mouse | |
Affinity Purified | |
Neuroscience | |
154043.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction