Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNKSR3 Antibody (OTI1D7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245630
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
CNKSR3 Monoclonal specifically detects CNKSR3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CNKSR3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500, Immunohistochemistry 1:150 | |
CNK homolog protein 3, CNK3, CNKSR family member 3MAGI1, FLJ31349, membrane associated guanylate kinase interacting protein-like 1, membrane-associated guanylate kinase-interacting protein-like 1, WW and PDZ domain containing 1 | |
Mouse | |
61.7 kDa | |
0.1 mL | |
Neuroscience | |
154043 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1D7 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
CNKSR3 | |
Full length human recombinant protein of human CNKSR3 (NP_775786) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction