Learn More
Invitrogen™ CNKSR3 Monoclonal Antibody (OTI1D7)

Description
CNKSR3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | CNKSR3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1D7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CNKSR3 |
| Gene Accession No. | Q6P9H4 |
| Gene Alias | 6820402C05; BC024086; CNK homolog protein 3; CNK3; CNKSR family member 3; Cnksr3; Connector enhancer of kinase suppressor of ras 3; connector enhancer of KSR 3; KOX14; MAGI1; maguin-like protein; membrane associated guanylate kinase interacting protein-like 1; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase-interacting protein-like 1; parturition-related protein 4; Prp4; ZNF21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.