Learn More
Invitrogen™ CNNM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120530
Description
Positive test controls include: Mouse brain, Rat brain. Immunogen sequence: NGAFTYYGVP AIMTTACSDN DVRKVGSLAG SSVFLNRSPS RCSGLNRSES PNRERSDFGG SNTQLYSSSN NLYMPDYSVH ILSDVQFVKI TRQQYQNALT ACHMDSSPQS PDMEAFTDGD STKAPTTRGT PQTPKDDPAI TLLNNRNSLP CSRSDGLRSP SEVVYLRMEE LAFTQEEMTD FEEHSTQQLT LSPAAVPTRA ASDSECCNIN LDTETSPCSS DFEENVGKKL LRTLSGQKRK RSPEGERTSE DNSNLTPLIT.
Probable metal transporter.
Specifications
CNNM1 | |
Polyclonal | |
Unconjugated | |
CNNM1 | |
ACDP1; AI846314; ancient conserved domain protein 1; ancient conserved domain-containing protein 1; CLP-1; Cnnm1; cyclin and CBS domain divalent metal cation transport mediator 1; cyclin M1; cyclin-like protein 1; Cyclin-M1; mACDP1; Metal transporter CNNM1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26507, 309387, 83674 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
2.48 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q0GA42, Q9NRU3 | |
CNNM1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 692-951 of human CNNM1 (NP_065081.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.