Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNNM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100862 Supplier Invitrogen™ Supplier No. PA5100862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CNNM1.

Probable metal transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNNM1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CNNM1
Gene Accession No. Q0GA42, Q9NRU3
Gene Alias ACDP1; AI846314; ancient conserved domain protein 1; ancient conserved domain-containing protein 1; CLP-1; Cnnm1; cyclin and CBS domain divalent metal cation transport mediator 1; cyclin M1; cyclin-like protein 1; Cyclin-M1; mACDP1; Metal transporter CNNM1
Gene Symbols CNNM1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CNNM1(Accession Q9NRU3), corresponding to amino acid residues E554-K604.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26507, 83674
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.