Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNNM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP315497100UL
This item is not returnable.
View return policy
Description
CNNM1 Polyclonal antibody specifically detects CNNM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CNNM1 | |
Polyclonal | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
ACDP1cyclin-M1, ancient conserved domain protein 1, Ancient conserved domain-containing protein 1, cyclin M1, Cyclin-M1, FLJ31632, metal transporter CNNM1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 692-951 of human CNNM1 (NP_065081.2). | |
100 μg | |
Cell Biology, Cell Cycle and Replication | |
26507 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction