Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNNM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566298 Supplier Invitrogen™ Supplier No. PA566298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVGENEETVII GLRLEDTNDV SFMEGGALRV SERTRVKLRV YGQNINNETW SRIAFTEHE Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

CNNM2 is a divalent metal cation transporter. CNNM2 mediates transport of divalent metal cations in an order of Mg2+ > Co2+ > Mn2+ > Sr2+ > Ba2+ > Cu2+ > Fe2+.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNNM2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNNM2
Gene Accession No. Q9H8M5
Gene Alias ACDP2; ancient conserved domain protein 2; ancient conserved domain-containing protein 2; AU015877; AW048635; Clp2; Cnnm2; cyclin and CBS domain divalent metal cation transport mediator 2; cyclin M2; cyclin-like protein 2; cyclin-M2; HOMG6; HOMGSMR; mACDP2; metal transporter CNNM2
Gene Symbols CNNM2
Host Species Rabbit
Immunogen Recombinant Human CNNM2. Recombinant protein control fragment (Product #RP-105143).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54805
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.