Learn More
Invitrogen™ CNNM3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total Cyclin M3 protein.
Specifications
Specifications
| Antigen | CNNM3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CNNM3 |
| Gene Accession No. | Q32NY4, Q8NE01 |
| Gene Alias | Acdp3; ancient conserved domain protein 3; ancient conserved domain-containing protein 3; Clp3; Cnnm3; cyclin and CBS domain divalent metal cation transport mediator 3; cyclin M3; cyclin-M3; mACDP3; metal transporter CNNM3 |
| Gene Symbols | CNNM3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.