Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNNM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114722
Description
Antibody detects endogenous levels of total CNNM3.
Probable metal transporter. [UniProt].
Specifications
CNNM3 | |
Polyclonal | |
Unconjugated | |
CNNM3 | |
Acdp3; ancient conserved domain protein 3; ancient conserved domain-containing protein 3; Clp3; Cnnm3; cyclin and CBS domain divalent metal cation transport mediator 3; cyclin M3; cyclin-M3; mACDP3; metal transporter CNNM3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26505 | |
-20°C | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q8NE01 | |
CNNM3 | |
A synthesized peptide derived from human CNNM3(Accession Q8NE01), corresponding to amino acid residues L271-L321. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction