Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNNM4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen CNNM4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159423
SDP
View Documents
Novus Biologicals
NBP159423
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

CNNM4 Polyclonal specifically detects CNNM4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CNNM4
Polyclonal
Rabbit
Vision
ACDP4cyclin-M4, Ancient conserved domain-containing protein 4, cyclin M4, Cyclin-M4, FLJ37746, FLJ42791, KIAA1592ancient conserved domain protein 4, metal transporter CNNM4
CNNM4
IgG
Western Blot
Unconjugated
RUO
Q6P4Q7
26504
Synthetic peptides corresponding to CNNM4(cyclin M4) The peptide sequence was selected from the middle region of CNNM4. Peptide sequence LLASRMENSPQFPIDGCTTHMENLAEKSELPVVDETTTLLNERNSLLHKA.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.