Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNNM4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | CNNM4 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CNNM4 Polyclonal specifically detects CNNM4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CNNM4 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
ACDP4cyclin-M4, Ancient conserved domain-containing protein 4, cyclin M4, Cyclin-M4, FLJ37746, FLJ42791, KIAA1592ancient conserved domain protein 4, metal transporter CNNM4 | |
CNNM4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Vision | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
26504 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FSYYGTMALTSVPSDRSPAHPTPLSRSASLSYPDRTDVSTAATLAGSSNQFGSSVLGQYISDFSVRALVDLQYIKITRQQYQNGLLASRMENSPQFPIDGCTTHMENLAEKSELPVVDETTTLLNERNSL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title