Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNNM4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 140661AP150UL
Description
This gene encodes a member of the ancient conserved domain containing protein family. Members of this protein family contain a cyclin box motif and have structural similarity to the cyclins. The encoded protein may play a role in metal ion transport. Mutations in this gene are associated with Jalili syndrome which consists of cone-rod dystrophy and amelogenesis imperfecta.Specifications
| CNNM4 | |
| Polyclonal | |
| Unconjugated | |
| CNNM4 | |
| ACDP4, CNNM4, cyclin M4, KIAA1592, Metal transporter CNNM4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26504, 363216, 94220 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P0C588, Q69ZF7, Q6P4Q7 | |
| Cnnm4 | |
| CNNM4 Fusion Protein Ag5121 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction