Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560534 Supplier Invitrogen™ Supplier No. PA560534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YAACLLPGAG ARPEVEALDA SLEDLLTRVD EFVGMLDMLR GDSSHVVSEG VPRIHAKAAE MRRIYSR Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.

The BLOC-1 complex is required for normal biogenesis of lysosome-related organelles, such as platelet dense granules and melanosomes. Plays a role in intracellular vesicle trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNO
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BLOC1S4
Gene Accession No. Q9NUP1
Gene Alias 2610101N07Rik; BCAS4L; biogenesis of lysosomal organelles complex 1 subunit 4; biogenesis of lysosomal organelles complex-1, subunit 4, cappuccino; biogenesis of lysosome-related organelles complex 1 subunit 4; biogenesis of organelles complex-1, subunit 4, cappuccino; BLOC-1 subunit 4; BLOC1S4; Blos4; cappuccino; cappuccino homolog; CNO; Protein cappuccino homolog; RGD1306037
Gene Symbols BLOC1S4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNO. Recombinant protein control fragment (Product #RP-99983).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55330
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.