Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNOT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | CNOT1 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CNOT1 Polyclonal specifically detects CNOT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CNOT1 | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
AD-005, adrenal gland protein AD-005, CCR4-associated factor 1, CCR4-NOT transcription complex subunit 1, CCR4-NOT transcription complex, subunit 1, CDC39DKFZp686E0722, hNOT1, KIAA1007DKFZp686O168, Negative regulator of transcription subunit 1 homolog, NOT1 (negative regulator of transcription 1, yeast) homolog, NOT1FLJ90644, NOT1HFLJ36492 | |
CNOT1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Rabbit | |
Human | |
A5YKK6 | |
23019 | |
This antibody was developed against a recombinant protein corresponding to amino acids: ALLQINTSWHTLRHELISTLMPIFLGNHPNSAIILHYAWHGQGQSPSIRQLIMHAMAEWYMRGEQYDQAKLSRILDVAQDLKALSMLLNGT | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title