Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNOT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562024
Description
Immunogen sequence: ALLQINTSWH TLRHELISTL MPIFLGNHPN SAIILHYAWH GQGQSPSIRQ LIMHAMAEWY MRGEQYDQAK LSRILDVAQD LKALSMLLNG T Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Scaffolding component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Its scaffolding function implies its interaction with the catalytic complex module and diverse RNA-binding proteins mediating the complex recruitment to selected mRNA 3'UTRs. Involved in degradation of AU-rich element (ARE)-containing mRNAs probably via association with ZFP36. Mediates the recruitment of the CCR4-NOT complex to miRNA targets and to the RISC complex via association with TNRC6A, TNRC6B or TNRC6C. Acts as a transcriptional repressor. Represses the ligand-dependent transcriptional activation by nuclear receptors. Involved in the maintenance of embryonic stem (ES) cell identity. [UniProt].
Specifications
CNOT1 | |
Polyclonal | |
Unconjugated | |
CNOT1 | |
6030411K04Rik; AA815922; AD-005; adrenal gland protein AD-005; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 1; CCR4-NOT transcription complex, subunit 1; CDC39; Cnot1; D830048B13; Kiaa1007; Negative regulator of transcription subunit 1 homolog; NOT1; NOT1 (negative regulator of transcription 1, yeast) homolog; NOT1H; RGD1308009 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23019 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A5YKK6 | |
CNOT1 | |
Recombinant protein corresponding to Human CNOT1. Recombinant protein control fragment (Product #RP-95505). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction