Learn More
CNOT1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305787AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this product is predicted to react with zebrafish, X. tropicalis
Scaffolding component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Its scaffolding function implies its interaction with the catalytic complex module and diverse RNA-binding proteins mediating the complex recruitment to selected mRNA 3′UTRs. Involved in degradation of AU-rich element (ARE)-containing mRNAs probably via association with ZFP36. Mediates the recruitment of the CCR4-NOT complex to miRNA targets and to the RISC complex via association with TNRC6A, TNRC6B or TNRC6C. Acts as a transcriptional repressor. Represses the ligand-dependent transcriptional activation by nuclear receptors. Involved in the maintece of emryonic stem (ES) cell identity.Specifications
CNOT1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
A5YKK6, Q6ZQ08 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23019, 234594 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
CNOT1 | |
Cnot1 | |
between 1400 and 1450 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.