Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNOT10 Antibody (PCRP-CNOT10-1D5), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314266MFV450
Description
CNOT10 Monoclonal antibody specifically detects CNOT10 in Human samples. It is validated for Flow CytometrySpecifications
CNOT10 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
25904 | |
Store at 4C in the dark. | |
IgG2c |
Flow Cytometry | |
PCRP-CNOT10-1D5 | |
Flow Cytometry | |
CCR4-NOT transcription complex subunit 10, CCR4-NOT transcription complex, subunit 10, DKFZp434K115, FLJ12890, FLJ13165 | |
Recombinant full-length human CNOT10 protein (Uniprot: Q9H9A5) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction