Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNOT10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100134
Description
Antibody detects endogenous levels of total CNOT10.
Component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Is not required for association of CNOT7 to the CCR4-NOT complex.
Specifications
CNOT10 | |
Polyclonal | |
Unconjugated | |
CNOT10 | |
2600001P13Rik; CCR4-NOT transcription complex subunit 10; CCR4-NOT transcription complex, subunit 10; Cnot10; FLJ12890; FLJ13165 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25904, 78893 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8BH15, Q9H9A5 | |
CNOT10 | |
A synthesized peptide derived from human CNOT10(Accession Q9H9A5), corresponding to amino acid residues L471-S521. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction