Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNOT10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557339 Supplier Invitrogen™ Supplier No. PA557339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNLMALNHAD KLLQQPKLSG SLKFLGHLYA AEALISLDRI SDAITHLNPE NVTDVSLGIS SNEQDQGSDK GENEAMESSG KRAPQCYPSS VNSARTV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Is not required for association of CNOT7 to the CCR4-NOT complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNOT10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNOT10
Gene Accession No. Q5XIA4, Q8BH15, Q9H9A5
Gene Alias 2600001P13Rik; CCR4-NOT transcription complex subunit 10; CCR4-NOT transcription complex, subunit 10; Cnot10; FLJ12890; FLJ13165
Gene Symbols CNOT10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CNOT10. Recombinant protein control fragment (Product #RP-96467).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25904, 316034, 78893
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.