Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNOT11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567078 Supplier Invitrogen™ Supplier No. PA567078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FRPEFIRPPPP LHICEDELAW LNPTEPDHAI QWDKSMCVKN STGVEIKRIM AKAFKSPLSS PQQTQLLGEL EKDPKLVYHI GLT Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

Component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Is required for the association of CNOT10 with the CCR4-NOT complex. Seems not to be required for complex deadenylase function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNOT11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNOT11
Gene Accession No. Q9UKZ1
Gene Alias C2orf29; C40; CCR4-NOT transcription complex subunit 11; CCR4-NOT transcription complex, subunit 11; CNOT11; UPF0760 protein C2orf29
Gene Symbols CNOT11
Host Species Rabbit
Immunogen Recombinant Human CNOT11. Recombinant protein control fragment (Product #RP-105489).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55571
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.