Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNOT2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130454AF350
Description
CNOT2 Polyclonal antibody specifically detects CNOT2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CNOT2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CCR4-associated factor 2, CCR4-NOT transcription complex, subunit 2, CDC36FLJ26456, negative regulator of transcription 2, NOT2CCR4-NOT transcription complex subunit 2, NOT2H | |
The immunogen for this product maps to a region between residue 490 and 540 of human CCR4-NOT transcription complex, subunit 2 using the numbering given in entry NP_055330.1 (GeneID 4848). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4848 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction