Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNOT2 Antibody (OTI5A12), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271202B
Description
CNOT2 Monoclonal specifically detects CNOT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CNOT2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CNOT2 | |
Full length recombinant protein of human CNOT2 (NP_055330) produced in E.coli. | |
0.1 mL | |
4848.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5A12 | |
Western Blot, Immunohistochemistry | |
CCR4-associated factor 2, CCR4-NOT transcription complex, subunit 2, CDC36FLJ26456, negative regulator of transcription 2, NOT2CCR4-NOT transcription complex subunit 2, NOT2H | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction