Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNOT3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572424
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2424 10 μL
50-157-2423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2424 Supplier Bethyl Laboratories, Inc Supplier No. IHC00623T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Dog, Orangutan, Monkey, Gorilla, Chimpanzee, Greater horseshoe bat, Small-eared galago and Gray s

The CCR4-NOT complex functions as general transcription regulation complex.

Specifications

Antigen CNOT3
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CNOT3
Gene Accession No. O75175
Gene Alias LENG2, NOT3, NOT3H, CCR4-NOT transcription complex subunit 3, CCR4-associated factor 3, NOT3 (negative regulator of transcription 3, yeast) homolog, leukocyte receptor cluster member 2
Gene Symbols CNOT3
Host Species Rabbit
Immunogen Between 525 and 575
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4849
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.